Gene Transcripts
- Gene Symbol
- C7orf49
- Entrez Gene ID
- 78996
- Gene Name
- chromosome 7 open reading frame 49
- Gene Aliases
- MRI
- Chromosome Location
- Chr. 7: 135092303 - 135170826 on Build GRCh38
- UniGene ID
- Hs.643113
- Species
- Homo sapiens
- Gene Symbol
- TMEM140
- Entrez Gene ID
- 55281
- Gene Name
- transmembrane protein 140
- Gene Aliases
- Chromosome Location
- Chr. 7: 135148014 - 135166215 on Build GRCh38
- UniGene ID
- Hs.521213
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BU683951.1 |
- |
1 |
73 |
|
View Full Product Details