Gene Transcripts

Gene Symbol
CENPM
Entrez Gene ID
79019
Gene Name
centromere protein M
Gene Aliases
C22orf18, CENP-M, PANE1
Chromosome Location
Chr. 22: 41927747 - 41947164 on Build GRCh38
UniGene ID
Hs.208912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002876.2 NP_001002876.1 5 819
NM_001110215.2 NP_001103685.1 2 502
NM_001304370.1 NP_001291299.1 5 1024
NM_001304372.1 NP_001291301.1 5 1109
NM_001304373.1 NP_001291302.1 4 932
NM_024053.4 NP_076958.1 6 911
GenBank mRNA AK123479.1 - 6 1936
AL160131.1 - 6 859
BC000705.2 - 6 858 3349507
BC007495.2 - 6 871 2966541
BX332140.2 - 4 868
BX417558.2 - 4 870
EU035297.1 - 2 502

View Full Product Details