Gene Transcripts
- Gene Symbol
- WDR25
- Entrez Gene ID
- 79446
- Gene Name
- WD repeat domain 25
- Gene Aliases
- C14orf67
- Chromosome Location
- Chr. 14: 100376410 - 100530303 on Build GRCh38
- UniGene ID
- Hs.497600
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161476.1 |
NP_001154948.1
|
3, 4 |
1071 |
|
NM_024515.4 |
NP_078791.3
|
3, 4 |
1202 |
|
XM_005268056.4 |
XP_005268113.1
|
3, 4 |
1078 |
|
XM_005268057.3 |
XP_005268114.1
|
2, 3 |
228 |
|
XM_005268058.4 |
XP_005268115.1
|
2, 3 |
242 |
|
XM_011537150.2 |
XP_011535452.1
|
3, 4 |
1325 |
|
XM_011537151.2 |
XP_011535453.2
|
2, 3 |
681 |
|
XM_017021639.1 |
XP_016877128.1
|
2, 3 |
230 |
|
GenBank mRNA |
AK291960.1 |
- |
3, 4 |
1034 |
|
AY295082.1 |
- |
3, 4 |
1085 |
|
BC003641.2 |
- |
2, 3 |
308 |
3529568 |
CT003969.1 |
- |
2, 3 |
234 |
|
View Full Product Details