Gene Transcripts

Gene Symbol
PYROXD1
Entrez Gene ID
79912
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 1
Gene Aliases
Chromosome Location
Chr. 12: 21437604 - 21471250 on Build GRCh38
UniGene ID
Hs.709545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024854.3 NP_079130.2 6 744
XM_006719153.3 XP_006719216.1 6 718
GenBank mRNA AK025681.1 - 6 646
AK125461.1 - 6 1157
AK293820.1 - 5 659
AK307710.1 - 5 569
AL832441.1 - 6 671
BC021662.1 - 6 645 3826938

View Full Product Details