Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077484.1 | NP_001070952.1 | 5 | 982 | |
NM_001278387.1 | NP_001265316.1 | 6 | 1444 | ||
NM_001278388.1 | NP_001265317.1 | 4 | 1206 | ||
NM_001278389.1 | NP_001265318.1 | 5 | 471 | ||
NM_001278390.1 | NP_001265319.1 | 5 | 1184 | ||
NM_030674.3 | NP_109599.3 | 5 | 1321 | ||
XM_011538784.2 | XP_011537086.1 | 5 | 706 | ||
XM_011538786.2 | XP_011537088.1 | 5 | 706 | ||
XM_011538787.2 | XP_011537089.1 | 5 | 706 | ||
XM_017019990.1 | XP_016875479.1 | 5 | 936 | ||
GenBank mRNA | AF247166.1 | - | 5 | 667 | |
AF271070.1 | - | 5 | 565 | ||
AK027825.1 | - | 5 | 471 | ||
AK074797.1 | - | 2 | 40 | ||
AK074851.1 | - | 3 | 182 | ||
AK074949.1 | - | 3 | 229 | ||
AK225639.1 | - | 5 | 559 | ||
AK308129.1 | - | 3 | 270 | ||
AK309410.1 | - | 5 | 898 | ||
AK313731.1 | - | 3 | 272 | ||
BC010620.1 | - | 5 | 528 | 3871101 | |
BX647547.1 | - | 6 | 1016 | ||
BX648495.1 | - | 5 | 909 | ||
BX648676.1 | - | 4 | 776 |