Gene Transcripts
- Gene Symbol
- HDHD2
- Entrez Gene ID
- 84064
- Gene Name
- haloacid dehalogenase like hydrolase domain containing 2
- Gene Aliases
- 3110052N05Rik, HEL-S-301
- Chromosome Location
- Chr. 18: 47107402 - 47150520 on Build GRCh38
- UniGene ID
- Hs.465041
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318765.1 |
NP_001305694.1
|
3, 4 |
508 |
|
NM_032124.4 |
NP_115500.1
|
3, 4 |
451 |
|
XM_005258371.4 |
XP_005258428.1
|
3, 4 |
471 |
|
XM_006722557.2 |
XP_006722620.1
|
4, 5 |
543 |
|
XM_011526227.2 |
XP_011524529.1
|
4, 5 |
527 |
|
XM_011526228.2 |
XP_011524530.1
|
4, 5 |
525 |
|
XM_017026039.1 |
XP_016881528.1
|
4, 5 |
564 |
|
XM_017026040.1 |
XP_016881529.1
|
4, 5 |
543 |
|
GenBank mRNA |
AK054590.1 |
- |
3, 4 |
488 |
|
AK292098.1 |
- |
3, 4 |
451 |
|
AL136681.1 |
- |
3, 4 |
442 |
|
BC011894.2 |
- |
3, 4 |
472 |
3944181 |
BC033031.1 |
- |
3, 4 |
487 |
4793566 |
BC038100.2 |
- |
3, 4 |
451 |
5266439 |
CR533487.1 |
- |
2, 3 |
318 |
|
FJ224322.1 |
- |
3, 4 |
442 |
|
View Full Product Details