Gene Transcripts
- Gene Symbol
- HPS3
- Entrez Gene ID
- 84343
- Gene Name
- HPS3, biogenesis of lysosomal organelles complex 2 subunit 1
- Gene Aliases
- BLOC2S1, SUTAL
- Chromosome Location
- Chr. 3: 149129584 - 149173196 on Build GRCh38
- UniGene ID
- Hs.558314
- Species
- Homo sapiens
- Gene Symbol
- CP
- Entrez Gene ID
- 1356
- Gene Name
- ceruloplasmin (ferroxidase)
- Gene Aliases
- CP-2
- Chromosome Location
- Chr. 3: 149162410 - 149222050 on Build GRCh38
- UniGene ID
- Hs.558314
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CB321657.1 |
- |
1 |
320 |
|
View Full Product Details