Gene Transcripts

Gene Symbol
PNMA6A
Entrez Gene ID
84968
Gene Name
paraneoplastic Ma antigen family member 6A
Gene Aliases
MA6, PNMA6, PNMA6C
Chromosome Location
Chr. X: 153072414 - 153075019 on Build GRCh38
UniGene ID
Hs.533301
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032882.5 NP_116271.3 2 1514
GenBank mRNA AK055322.1 - 2 1446
AK289879.1 - 2 1492
BC007631.1 - 1 1246 3507248

View Full Product Details