Gene Transcripts

Gene Symbol
RBM18
Entrez Gene ID
92400
Gene Name
RNA binding motif protein 18
Gene Aliases
Chromosome Location
Chr. 9: 122239555 - 122264864 on Build GRCh38
UniGene ID
Hs.415842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033117.3 NP_149108.1 2 240
NR_027125.1 2 240
NR_027126.1 2 240
XM_006717319.1 XP_006717382.1 2 196
GenBank mRNA AK057676.1 - 2 216
AK299625.1 - 2 216
AK301050.1 - 2 196
AK310008.1 - 2 197
AK310807.1 - 2 182
AL389986.1 - 2 168
BC008942.2 - 2 187 2822634
BC019319.1 - 2 187 2822634
BP870502.1 - 2 219
DA308339.1 - 2 240

View Full Product Details