Gene Transcripts

Gene Symbol
C5orf30
Entrez Gene ID
90355
Gene Name
chromosome 5 open reading frame 30
Gene Aliases
Chromosome Location
Chr. 5: 103258702 - 103278660 on Build GRCh38
UniGene ID
Hs.482976
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316968.1 NP_001303897.1 3 704
NM_001316969.1 NP_001303898.1 3 420
NM_033211.3 NP_149988.1 3 464
GenBank mRNA AF038182.1 - 3 358
AK058020.1 - 3 400
AK090558.1 - 3 636
BC009203.2 - 3 425 3628991
BG722102.1 - 3 474
DA503423.1 - 3 420

View Full Product Details