Gene Transcripts

Gene Symbol
SLC5A11
Entrez Gene ID
115584
Gene Name
solute carrier family 5 member 11
Gene Aliases
KST1, RKST1, SGLT6, SMIT2
Chromosome Location
Chr. 16: 24832784 - 24911711 on Build GRCh38
UniGene ID
Hs.164118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258411.2 NP_001245340.1 9 975
NM_001258412.2 NP_001245341.1 8 870
NM_001258413.2 NP_001245342.1 9 921
NM_052944.4 NP_443176.2 9 931
XM_005255080.2 XP_005255137.1 10 959
XM_005255082.4 XP_005255139.1 4 365
XM_006721015.2 XP_006721078.1 8 939
XM_011545725.2 XP_011544027.2 9 1102
XM_017022900.1 XP_016878389.1 10 1275
XM_017022901.1 XP_016878390.1 10 1985
XM_017022902.1 XP_016878391.1 10 1114
XM_017022903.1 XP_016878392.1 11 1124
XM_017022904.1 XP_016878393.1 11 1294
XM_017022905.1 XP_016878394.1 9 1170
XM_017022906.1 XP_016878395.1 10 1135
GenBank mRNA AF292385.1 - 8 699
AJ305237.1 - 8 717
AK125267.1 - 11 1047
AK295427.1 - 8 739
AK298373.1 - 8 716
AK303691.1 - 9 843
AY044906.1 - 9 931
BC057780.1 - 9 790 5287366
U41898.1 - 6 483

View Full Product Details