Gene Transcripts
- Gene Symbol
- KLHDC3
- Entrez Gene ID
- 116138
- Gene Name
- kelch domain containing 3
- Gene Aliases
- PEAS, dJ20C7.3
- Chromosome Location
- Chr. 6: 43014103 - 43021298 on Build GRCh38
- UniGene ID
- Hs.412468
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_057161.3 |
NP_476502.1
|
4, 5 |
760 |
|
GenBank mRNA |
AB055925.1 |
- |
4, 5 |
624 |
|
AK097719.1 |
- |
2, 3 |
486 |
|
AK290384.1 |
- |
4, 5 |
616 |
|
AK302336.1 |
- |
3, 4 |
399 |
|
AK308917.1 |
- |
4, 5 |
646 |
|
BC000295.2 |
- |
4, 5 |
580 |
2819570 |
BC001789.1 |
- |
4, 5 |
595 |
3355470 |
BC001793.2 |
- |
4, 5 |
558 |
3352397 |
BC007296.1 |
- |
4, 5 |
610 |
3349731 |
BC009460.1 |
- |
4, 5 |
608 |
3537223 |
BC021546.1 |
- |
4, 5 |
597 |
3351635 |
View Full Product Details