Gene Transcripts

Gene Symbol
C2orf15
Entrez Gene ID
150590
Gene Name
chromosome 2 open reading frame 15
Gene Aliases
Chromosome Location
Chr. 2: 99141613 - 99151487 on Build GRCh38
UniGene ID
Hs.352211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317992.1 NP_001304921.1 5 796
NM_144706.3 NP_653307.1 4 700
GenBank mRNA BC021264.2 - 4 591

View Full Product Details