Gene Transcripts

Gene Symbol
C3orf30
Entrez Gene ID
152405
Gene Name
chromosome 3 open reading frame 30
Gene Aliases
TSCPA
Chromosome Location
Chr. 3: 119146150 - 119151455 on Build GRCh38
UniGene ID
Hs.730099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152539.2 NP_689752.2 1 138
GenBank mRNA AK057421.1 - 1 138
BC130475.1 - 1 137 40146506
BC144197.1 - 1 137 9052717

View Full Product Details