Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304949.1 | NP_001291878.1 | 5 | 910 | |
NM_001304951.1 | NP_001291880.1 | 2 | 599 | ||
NM_001304952.1 | NP_001291881.1 | 5 | 831 | ||
NM_001304953.1 | NP_001291882.1 | 5 | 1129 | ||
NM_001304954.1 | NP_001291883.1 | 4 | 928 | ||
NM_152465.2 | NP_689678.1 | 4 | 864 | ||
XM_005257927.4 | XP_005257984.1 | 5 | 1100 | ||
XM_006721723.3 | XP_006721786.1 | 5 | 1097 | ||
XM_011524398.2 | XP_011522700.1 | 5 | 830 | ||
XM_011524399.2 | XP_011522701.1 | 4 | 745 | ||
XM_011524400.2 | XP_011522702.1 | 4 | 654 | ||
XM_011524401.1 | XP_011522703.1 | 3 | 655 | ||
XM_017024237.1 | XP_016879726.1 | 4 | 748 | ||
GenBank mRNA | AK301868.1 | - | 5 | 828 | |
BC029574.1 | - | 4 | 811 | 5267334 | |
BQ215318.1 | - | 2 | 439 | ||
CR984201.1 | - | 5 | 590 |