Gene Transcripts
- Gene Symbol
- KCNRG
- Entrez Gene ID
- 283518
- Gene Name
- potassium channel regulator
- Gene Aliases
- DLTET
- Chromosome Location
- Chr. 13: 50015254 - 50020922 on Build GRCh38
- UniGene ID
- Hs.660161
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173605.1 |
NP_775876.1
|
1, 2 |
826 |
|
NM_199464.2 |
NP_955751.1
|
2, 3 |
924 |
|
GenBank mRNA |
AK313401.1 |
- |
2, 3 |
775 |
|
AY129653.1 |
- |
2, 3 |
734 |
|
AY129654.1 |
- |
1, 2 |
644 |
|
AY169387.1 |
- |
2, 3 |
924 |
|
AY169388.1 |
- |
1, 2 |
826 |
|
AY190921.1 |
- |
1, 2 |
606 |
|
AY190922.1 |
- |
2, 3 |
704 |
|
BC020887.1 |
- |
2, 3 |
734 |
4690662 |
BC032343.1 |
- |
1, 2 |
620 |
5207702 |
View Full Product Details