Gene Transcripts
- Gene Symbol
- HIST2H2AB
- Entrez Gene ID
- 317772
- Gene Name
- histone cluster 2, H2ab
- Gene Aliases
- H2AB
- Chromosome Location
- Chr. 1: 149887469 - 149887916 on Build GRCh38
- UniGene ID
- Hs.664173
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175065.2 |
NP_778235.1
|
1 |
274 |
|
View Full Product Details