Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001946.3 | NP_001937.2 | 3 | 2171 | |
NM_022652.3 | NP_073143.2 | 2 | 1733 | ||
GenBank mRNA | AB013382.1 | - | 3 | 1702 | |
AB013602.1 | - | 2 | 1264 | ||
AK091753.1 | - | 4 | 1788 | ||
AK222882.1 | - | 2 | 1279 | ||
BC003143.1 | - | 3 | 1699 | 2906126 | |
BC003562.1 | - | 3 | 1699 | 3547162 | |
BC005047.1 | - | 3 | 1775 | 3954486 | |
BC037236.1 | - | 3 | 1798 | 5183133 | |
X93920.1 | - | 3 | 1702 |