Gene Transcripts

Gene Symbol
NKX2-8
Entrez Gene ID
26257
Gene Name
NK2 homeobox 8
Gene Aliases
NKX2.8, NKX2H, Nkx2-9
Chromosome Location
Chr. 14: 36580011 - 36582614 on Build GRCh38
UniGene ID
Hs.234763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014360.3 NP_055175.2 2 620
GenBank mRNA AF000295.1 - 2 594
AK313537.1 - 2 613
BC041090.1 - 2 587 5752253

View Full Product Details