Gene Transcripts

Gene Symbol
HOXB5
Entrez Gene ID
3215
Gene Name
homeobox B5
Gene Aliases
HHO.C10, HOX2, HOX2A, HU-1, Hox2.1
Chromosome Location
Chr. 17: 48591257 - 48593741 on Build GRCh38
UniGene ID
Hs.654456
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002147.3 NP_002138.1 2 1460
GenBank mRNA BC004454.1 - 1 389
BC008940.2 - 1 389
M92299.1 - 2 1677
X03794.1 - 1 811

View Full Product Details