Gene Transcripts

Gene Symbol
MRGPRX3
Entrez Gene ID
117195
Gene Name
MAS related GPR family member X3
Gene Aliases
GPCR, MRGX3, SNSR1, SNSR2
Chromosome Location
Chr. 11: 18120955 - 18138480 on Build GRCh38
UniGene ID
Hs.380177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054031.3 NP_473372.3 3 665
XM_011519882.2 XP_011518184.1 2 520
GenBank mRNA BC067292.1 - 3 665
EU432126.1 - 1 304

View Full Product Details