Gene Transcripts

Gene Symbol
HIST1H4C
Entrez Gene ID
8364
Gene Name
histone cluster 1, H4c
Gene Aliases
H4/g, H4FG, dJ221C16.1
Chromosome Location
Chr. 6: 26103948 - 26104337 on Build GRCh38
UniGene ID
Hs.46423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003542.3 NP_003533.1 1 143
GenBank mRNA BC054014.1 - 1 199
BC130558.1 - 1 215 40146589
BC130560.1 - 1 215 40146591

View Full Product Details