Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003684.1 | NP_001003684.1 | 2 | 358 | |
NM_013387.3 | NP_037519.2 | 2 | 299 | ||
GenBank mRNA | AB028598.1 | - | 2 | 300 | |
AF112217.1 | - | 2 | 288 | ||
AF161468.1 | - | 2 | 286 | ||
AF161500.1 | - | 2 | 356 | ||
AF161536.1 | - | 2 | 305 | ||
AK225973.1 | - | 2 | 284 | ||
BC005402.1 | - | 2 | 299 | 3961930 | |
BC015971.1 | - | 2 | 293 | 4097106 | |
BI754905.1 | - | 2 | 261 | ||
BU935406.1 | - | 358 | |||
CB999839.1 | - | 2 | 301 | ||
CD674029.1 | - | 2 | 166 |