Gene Transcripts

Gene Symbol
Timm10b
Entrez Gene ID
84384
Gene Name
translocase of inner mitochondrial membrane 10 homolog B (yeast)
Gene Aliases
Fxc1, Tim9b
Chromosome Location
Chr. 1: 170471265 - 170474111 on Build Rnor_6.0
UniGene ID
Rn.13451
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053371.2 NP_445823.1 507
GenBank mRNA AF150106.1 - 508

View Full Product Details