Gene Transcripts

Gene Symbol
Brinp3
Entrez Gene ID
286901
Gene Name
bone morphogenetic protein/retinoic acid inducible neural-specific 3
Gene Aliases
Fam5c
Chromosome Location
Chr. 13: 63526068 - 63959390 on Build Rnor_6.0
UniGene ID
Rn.64406
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173121.1 NP_775144.1 2, 3 668
XM_006249965.2 XP_006250027.1 2, 3 1086
GenBank mRNA AB077854.1 - 2, 3 668

View Full Product Details