Gene Transcripts

Gene Symbol
FZD7
Entrez Gene ID
8324
Gene Name
frizzled class receptor 7
Gene Aliases
FzE3
Chromosome Location
Chr. 2: 202034587 - 202038437 on Build GRCh38
UniGene ID
Hs.173859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003507.1 NP_003498.1 1 342
GenBank mRNA AB010881.1 - 1 296
AB017365.1 - 1 342
BC015915.1 - 1 295 4549389

View Full Product Details