Gene Transcripts

Gene Symbol
Fnta
Entrez Gene ID
25318
Gene Name
farnesyltransferase, CAAX box, alpha
Gene Aliases
PFAS
Chromosome Location
Chr. 16: 70834957 - 70854724 on Build Rnor_6.0
UniGene ID
Rn.5990
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012847.1 NP_036979.1 4 544
GenBank mRNA BC085758.1 - 4 494
M81225.1 - 4 544

View Full Product Details