Gene Transcripts

Gene Symbol
Mak
Entrez Gene ID
25677
Gene Name
male germ cell-associated kinase
Gene Aliases
Chromosome Location
Chr. 17: 21531651 - 21567742 on Build Rnor_6.0
UniGene ID
Rn.9670
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013136.1 NP_037268.1 1, 2 345
GenBank mRNA BC078887.1 - 1, 2 333
M35862.1 - 1, 2 345

View Full Product Details