Gene Transcripts

Gene Symbol
Fabp6
Entrez Gene ID
25440
Gene Name
fatty acid binding protein 6, ileal
Gene Aliases
IBABP
Chromosome Location
Chr. 10: 29191621 - 29196292 on Build Rnor_6.0
UniGene ID
Rn.10008
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017098.1 NP_058794.1 2, 3 299
XM_008767638.1 XP_008765860.1 1, 2 371
GenBank mRNA L22788.1 - 2, 3 299
S52878.1 - 2, 3 278

View Full Product Details