Gene Transcripts

Gene Symbol
Spink13
Entrez Gene ID
100038417
Gene Name
serine peptidase inhibitor, Kazal type 13
Gene Aliases
ENSMUSG00000073551, Gm10534, Spink5l3
Chromosome Location
Chr. 18: 62605898 - 62741387 on Build GRCm38
UniGene ID
Mm.387557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168423.2 NP_001161895.1 5 576
GenBank mRNA AK139149.1 - 5 577

View Full Product Details