Gene Transcripts

Gene Symbol
Micalcl
Entrez Gene ID
100504195
Gene Name
MICAL C-terminal like
Gene Aliases
4921517J23Rik
Chromosome Location
Chr. 7: 112368308 - 112413104 on Build GRCm38
UniGene ID
Mm.291129
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027587.2 NP_081863.2 10 2234
GenBank mRNA AB359922.1 - 10 2234
AK014911.1 - 7 1672

View Full Product Details