Gene Transcripts

Gene Symbol
Slc5a10
Entrez Gene ID
109342
Gene Name
solute carrier family 5 (sodium/glucose cotransporter), member 10
Gene Aliases
C330021F16Rik, SGLT5
Chromosome Location
Chr. 11: 61672782 - 61720811 on Build GRCm38
UniGene ID
Mm.441070
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033227.2 NP_001028399.1 16 2192
GenBank mRNA AK082798.1 - 5 848
AK144033.1 - 16 2193
AK165337.1 - 15 1988

View Full Product Details