Gene Transcripts

Gene Symbol
Mcpt9
Entrez Gene ID
17232
Gene Name
mast cell protease 9
Gene Aliases
MMCP-1, MMCP-9, MMCP9
Chromosome Location
Chr. 14: 56026758 - 56030495 on Build GRCm38
UniGene ID
Mm.436621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010782.3 NP_034912.3 1 888
GenBank mRNA AY007568.1 - 1 888

View Full Product Details