Gene Transcripts

Gene Symbol
Wfikkn1
Entrez Gene ID
215001
Gene Name
WAP, FS, Ig, KU, and NTR-containing protein 1
Gene Aliases
Gasp2, Wfikkn
Chromosome Location
Chr. 17: 25877628 - 25880858 on Build GRCm38
UniGene ID
Mm.328871
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100454.1 NP_001093924.1 2 1018
GenBank mRNA BC048715.1 - 2 456
BC147298.1 - 2 236 9056349
BC147299.1 - 2 236 9056355

View Full Product Details