Gene Transcripts

Gene Symbol
Lclat1
Entrez Gene ID
225010
Gene Name
lysocardiolipin acyltransferase 1
Gene Aliases
AI181996, Agpat8, Alcat1, Gm91, Lycat
Chromosome Location
Chr. 17: 73107985 - 73243368 on Build GRCm38
UniGene ID
Mm.329194
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081071.2 NP_001074540.1 6 1927
NM_001177967.1 NP_001171438.1 6 1906
NM_001177968.1 NP_001171439.1 6 1895

View Full Product Details