Gene Transcripts

Gene Symbol
Slc27a6
Entrez Gene ID
225579
Gene Name
solute carrier family 27 (fatty acid transporter), member 6
Gene Aliases
4732438L20Rik, FACVL2, FATP6, VLCS-H1
Chromosome Location
Chr. 18: 58556240 - 58612869 on Build GRCm38
UniGene ID
Mm.258517
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081072.1 NP_001074541.1 10 2389
GenBank mRNA AK028699.1 - 10 2391
BC115435.1 - 10 2372 40094587

View Full Product Details