Gene Transcripts

Gene Symbol
Cym
Entrez Gene ID
229697
Gene Name
chymosin
Gene Aliases
Gm131
Chromosome Location
Chr. 3: 107211295 - 107221732 on Build GRCm38
UniGene ID
Mm.297051
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111143.1 NP_001104613.1 5 612
GenBank mRNA BC147477.1 - 5 609
BC171994.1 - 5 609 9054280

View Full Product Details