Gene Transcripts

Gene Symbol
Ogdhl
Entrez Gene ID
239017
Gene Name
oxoglutarate dehydrogenase-like
Gene Aliases
Chromosome Location
Chr. 14: 32322012 - 32348150 on Build GRCm38
UniGene ID
Mm.318302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081130.1 NP_001074599.1 23 2994
GenBank mRNA BC157970.1 - 22 2794
BC157971.1 - 22 2794 9007293

View Full Product Details