Gene Transcripts

Gene Symbol
Dytn
Entrez Gene ID
241073
Gene Name
dystrotelin
Gene Aliases
Gm215
Chromosome Location
Chr. 1: 63622851 - 63686927 on Build GRCm38
UniGene ID
Mm.297680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081658.1 NP_001075127.1 14 1773
GenBank mRNA DQ443727.1 - 14 1773

View Full Product Details