Gene Transcripts
- Gene Symbol
- Ptprj
- Entrez Gene ID
- 29645
- Gene Name
- protein tyrosine phosphatase, receptor type, J
- Gene Aliases
- DEP-1
- Chromosome Location
- Chr. 3: 79233519 - 79390956 on Build Rnor_6.0
- UniGene ID
- Rn.10278
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017269.2 |
NP_058965.2
|
14, 15 |
2932 |
|
GenBank mRNA |
U40790.1 |
- |
14, 15 |
2790 |
|
View Full Product Details