Gene Transcripts

Gene Symbol
Dpcr1
Entrez Gene ID
268949
Gene Name
diffuse panbronchiolitis critical region 1 (human)
Gene Aliases
Gm630
Chromosome Location
Chr. 17: 35635755 - 35643695 on Build GRCm38
UniGene ID
Mm.296301
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033366.3 NP_001028538.1 3 1592
GenBank mRNA AK131887.1 - 3 1594
AK164929.1 - 3 1593

View Full Product Details