Gene Transcripts

Gene Symbol
Prps1l3
Entrez Gene ID
328099
Gene Name
phosphoribosyl pyrophosphate synthetase 1-like 3
Gene Aliases
AU021838, AW536152, Gm5081
Chromosome Location
Chr. 12: 57230412 - 57242168 on Build GRCm38
UniGene ID
Mm.268180
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037746.3 NP_001032835.2 2 3633
GenBank mRNA AK089022.1 - 1 662

View Full Product Details