Gene Transcripts

Gene Symbol
Mast4
Entrez Gene ID
328329
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
4930420O11Rik, AI642422, mKIAA0303
Chromosome Location
Chr. 13: 102732488 - 103335840 on Build GRCm38
UniGene ID
Mm.202606
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175171.3 NP_780380.2 29 10261
GenBank mRNA AK090136.1 - 29 10246
AK129114.1 - 1 4963
AK137360.1 - 1 4033
BC042511.1 - 16 4036

View Full Product Details