Gene Transcripts

Gene Symbol
Tmem92
Entrez Gene ID
544806
Gene Name
transmembrane protein 92
Gene Aliases
AU016430, Tmem92-ps
Chromosome Location
Chr. 11: 94777217 - 94782703 on Build GRCm38
UniGene ID
Mm.379774
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034896.2 NP_001030068.1 5 1843
NM_001163172.1 NP_001156644.1 5 1987
GenBank mRNA AK139848.1 - 5 1843

View Full Product Details