Gene Transcripts

Gene Symbol
Arsi
Entrez Gene ID
545260
Gene Name
arylsulfatase i
Gene Aliases
BC020108
Chromosome Location
Chr. 18: 60912240 - 60919504 on Build GRCm38
UniGene ID
Mm.20147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038499.1 NP_001033588.1 2 778
GenBank mRNA BC138970.1 - 2 824
BC141169.1 - 2 824 9055781

View Full Product Details