Gene Transcripts

Gene Symbol
Smok3a
Entrez Gene ID
545814
Gene Name
sperm motility kinase 3A
Gene Aliases
Smok2b1
Chromosome Location
Chr. 5: 138021276 - 138034665 on Build GRCm38
UniGene ID
Mm.349005
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168602.1 NP_001162073.1 2 2413
GenBank mRNA AJ245454.1 - 2 2413

View Full Product Details