Gene Transcripts

Gene Symbol
Otogl
Entrez Gene ID
628870
Gene Name
otogelin-like
Gene Aliases
EG628870, Gm6924
Chromosome Location
Chr. 10: 107760530 - 107912174 on Build GRCm38
UniGene ID
Mm.336090
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177567.1 NP_001171038.1 32 3725
GenBank mRNA GU140005.1 - 32 3726

View Full Product Details