Gene Transcripts

Gene Symbol
Vmn2r28
Entrez Gene ID
665255
Gene Name
vomeronasal 2, receptor 28
Gene Aliases
EG665255
Chromosome Location
Chr. 7: 5480456 - 5493851 on Build GRCm38
UniGene ID
Mm.425373
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081405.1 NP_001074874.1 3 777

View Full Product Details