Gene Transcripts

Gene Symbol
Slc25a53
Entrez Gene ID
67062
Gene Name
solute carrier family 25, member 53
Gene Aliases
2310046F18Rik, 2810402A17Rik, AI448995, AI841128, Mcart6
Chromosome Location
Chr. X: 136981116 - 137038302 on Build GRCm38
UniGene ID
Mm.276518
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082412.2 NP_001075881.1 7 2424
NM_001114179.1 NP_001107651.1 2 1806
GenBank mRNA AK076279.1 - 8 2392

View Full Product Details