Gene Transcripts

Gene Symbol
Arl16
Entrez Gene ID
70317
Gene Name
ADP-ribosylation factor-like 16
Gene Aliases
2600005N12Rik
Chromosome Location
Chr. 11: 120464326 - 120467600 on Build GRCm38
UniGene ID
Mm.485046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197995.2 NP_932112.2 5 1748
GenBank mRNA AK140077.1 - 3 1914

View Full Product Details